Clotrimazole
Clotrimazole
Clotrimazole
Thermo Scientific Chemicals
Clotrimazole
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ACR455460250
Price (MYR)
1,636.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,636.00
Add to cart
Chemical Identifiers
CAS1191-25-9
IUPAC Name6-hydroxyhexanoate
Molecular FormulaC6H11O3
InChI KeyIWHLYPDWHHPVAA-UHFFFAOYSA-M
SMILESOCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or liquid as melt
Assay (Silylated GC)≥94.0% (monomer + dimer)
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to white
Assay (Silylated GC)Monomer content: ≥80%