Thiamphenicol, 98%, Thermo Scientific Chemicals
Thiamphenicol, 98%, Thermo Scientific Chemicals
Thiamphenicol, 98%, Thermo Scientific Chemicals
Thiamphenicol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiamphenicol, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR455450010
Price (MYR)
417.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
417.00
Add to cart
Specifications
Chemical Name or MaterialThiamphenicol
CAS15318-45-3
EINECS Number239-355-3
Infrared SpectrumAuthentic
Assay Percent Range97.5% min. (HPLC)