Check Order Status
Teicoplanin, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
Teicoplanin, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
ACR455400010
1 g
Catalog number
ACR455400010
Price (MYR)
3,570.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
3,570.00
Add to cart
Specifications
Chemical Name or Material
Teicoplanin
Type
Teicoplanin
Physical Form
Powder
CAS
61036-62-2
Melting Point
260.0°C