3'-Azido-3'-deoxythymidine, 98%, Thermo Scientific Chemicals
3'-Azido-3'-deoxythymidine, 98%, Thermo Scientific Chemicals
3'-Azido-3'-deoxythymidine, 98%, Thermo Scientific Chemicals
3'-Azido-3'-deoxythymidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3'-Azido-3'-deoxythymidine, 98%, Thermo Scientific Chemicals
CAS: 30516-87-1 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR454980010
Price (MYR)
551.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
551.00
Add to cart
Chemical Identifiers
CAS13616-82-5
IUPAC Name2,4-dimethylbenzene-1-thiol
Molecular FormulaC8H10S
InChI KeyAMNLXDDJGGTIPL-UHFFFAOYSA-N
SMILESCC1=CC=C(S)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5670-1.5730 @ 20°C