Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
CAS: 541-02-6 | C10H30O5Si5 | 370.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ACR454580500
Price (MYR)
470.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
470.00
Add to cart
Chemical Identifiers
CAS111-68-2
IUPAC Nameheptan-1-amine
Molecular FormulaC7H17N
InChI KeyWJYIASZWHGOTOU-UHFFFAOYSA-N
SMILESCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.3 %
Infrared spectrumConforms
Refractive index1.4233 to 1.4253 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow