Barium nitrate, pure, Thermo Scientific Chemicals
Barium nitrate, pure, Thermo Scientific Chemicals
Barium nitrate, pure, Thermo Scientific Chemicals
Barium nitrate, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Barium nitrate, pure, Thermo Scientific Chemicals
CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number ACR453805000
Price (MYR)
432.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
432.00
Add to cart
Specifications
Chemical Name or MaterialBarium nitrate
Melting Point592.0°C
CAS10022-31-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
May intensify fire; oxidizer.