Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ACR453171000
Price (MYR)
945.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
945.00
Add to cart
Chemical Identifiers
CAS7773-06-0
IUPAC Nameammonium sulfamate
Molecular FormulaH6N2O3S
InChI KeyGEHMBYLTCISYNY-UHFFFAOYSA-N
SMILES[NH4+].NS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Melting point132.0°C to 134.0°C
Titration>=98.0 %
Heavy metals (as Pb)=<5 ppm