Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number ACR453170250
Price (MYR)
550.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
550.00
Add to cart
Chemical Identifiers
CAS2432-51-1
IUPAC Name1-(methylsulfanyl)butan-1-one
Molecular FormulaC5H10OS
InChI KeyGRLJIIJNZJVMGP-UHFFFAOYSA-N
SMILESCCCC(=O)SC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4605-1.4645 @ 20?C
Appearance (Color)Clear colorless