Itraconazole, 99%
Itraconazole, 99%
Itraconazole, 99%
Thermo Scientific Chemicals
Itraconazole, 99%
Itraconazole, or voriconazole, CAS # 84625-61-6, is an N-arylpiperazine that has antifungal properties.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ACR452870250
Price (MYR)
2,160.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,160.00
Add to cart
Chemical Identifiers
CAS10036-64-3
IUPAC NameN-[(2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular FormulaC8H15NO6
InChI KeyOVRNDRQMDRJTHS-PSLNIYNBNA-N
SMILESCC(=O)N[C@H]1[C@@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=99.0 %
Loss on drying=<1.0 % (105°C, 4 h)
View more
General Description
• Itraconazole is an aromatic ether that works as a strong inhibitor of P-glycoproteins and CYP3A4
• Itraconazole also acts as a hedgehog signaling inhibitor and P450 inhibitor
Applications
• Itraconazole can be exhibits antifungal activity against organisms including aspergillosis, blastomycosis, candidiasis, chromoblastomycosis, coccidioidomycosis, cryptococcosis, histoplasmosis, and sporotrichosis
• Itraconazole may be used to experimentally target the delivery of other molecules, such as plasmids, to cells
RUO – Research Use Only
General References:
  1. Sharma, P.; Bhalla, M.; Thami, G.P.; Chander, J. Evaluation of efficacy and safety of oral terbinafine and itraconazole combination therapy in the management of dermatophytosis. J Dermatolog Treat. 2020, 31(7), 749-753.
  2. Hamed Khater, M.; Khattab, F.M. Combined long-pulsed Nd-Yag laser and itraconazole versus itraconazole alone in the treatment of onychomycosis nails. J Dermatolog Treat. 2020, 31(4), 406-409.