Itraconazole, 99%
Itraconazole, 99%
Itraconazole, 99%
Thermo Scientific Chemicals
Itraconazole, 99%
Itraconazole, or voriconazole, CAS # 84625-61-6, is an N-arylpiperazine that has antifungal properties.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR452870050
Price (MYR)
734.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
734.00
Add to cart
Chemical Identifiers
CAS23132-21-0
IUPAC Name2-bromo-3-methyl-5-nitropyridine
Molecular FormulaC6H5BrN2O2
InChI KeyFZIQHPKXSLHGBZ-UHFFFAOYSA-N
SMILESCC1=CC(=CN=C1Br)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream to cream or pale yellow
Assay (GC)≥96.0%
Melting Point (clear melt)52.5-61.5?C
General Description
• Itraconazole is an aromatic ether that works as a strong inhibitor of P-glycoproteins and CYP3A4
• Itraconazole also acts as a hedgehog signaling inhibitor and P450 inhibitor
Applications
• Itraconazole can be exhibits antifungal activity against organisms including aspergillosis, blastomycosis, candidiasis, chromoblastomycosis, coccidioidomycosis, cryptococcosis, histoplasmosis, and sporotrichosis
• Itraconazole may be used to experimentally target the delivery of other molecules, such as plasmids, to cells
RUO – Research Use Only
General References:
  1. Sharma, P.; Bhalla, M.; Thami, G.P.; Chander, J. Evaluation of efficacy and safety of oral terbinafine and itraconazole combination therapy in the management of dermatophytosis. J Dermatolog Treat. 2020, 31(7), 749-753.
  2. Hamed Khater, M.; Khattab, F.M. Combined long-pulsed Nd-Yag laser and itraconazole versus itraconazole alone in the treatment of onychomycosis nails. J Dermatolog Treat. 2020, 31(4), 406-409.