1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
CAS: 84322-56-5 | C6F3I3 | 509.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR452690010
Price (MYR)
1,018.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,018.00
Add to cart
Chemical Identifiers
CAS3389-21-7
IUPAC Name3-(2-bromoethyl)-1H-indole
Molecular FormulaC10H10BrN
InChI KeyNTLAICDKHHQUGC-UHFFFAOYSA-N
SMILESBrCCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light brown
Infrared spectrumConforms
Melting point95°C to 101°C
Appearance (Form)Crystalline powder
NMR (1H-NMR)Consistent with structure