3-Methyl-1-butanol, for analysis
3-Methyl-1-butanol, for analysis
3-Methyl-1-butanol, for analysis
3-Methyl-1-butanol, for analysis
Acros Organics
3-Methyl-1-butanol, for analysis
CAS: 123-51-3 | C5H12O | 88.15 g/mol
Have Questions?
Catalog NumberQuantity
ACR4516600101 L
Catalog number ACR451660010
Price (MYR)
190.90
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
190.90
Add to cart
Specifications
Chemical Name or Material3-Methyl-1-butanol
CAS123-51-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin