Yeast extract
Thermo Scientific Chemicals
Yeast extract
CAS: 84604-16-
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number ACR451120010
Price (MYR)
814.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
814.00
Add to cart
Specifications
CAS84604-16-
Chemical Name or MaterialYeast Extract
EINECS Number283-294-5
FormatPlastic Bottle
Quantity1 kg