Sulfuric acid, for analysis, 25% solution in water
Sulfuric acid, for analysis, 25% solution in water
Sulfuric acid, for analysis, 25% solution in water
Sulfuric acid, for analysis, 25% solution in water
Thermo Scientific Chemicals
Sulfuric acid, for analysis, 25% solution in water
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
ACR4510500101 L
Catalog number ACR451050010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Length2.8 - 3.5mm
Total Metal Impurities0.02% max.
Appearance (Color)Gray
Diameter2.8 - 3.5mm
FormSlug