Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Thermo Scientific Chemicals
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
ACR4507200101 L
Catalog number ACR450720010
Price (MYR)
665.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
665.00
Add to cart
Chemical Identifiers
CAS569-58-4
IUPAC Nametriammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular FormulaC22H25N3O9
InChI KeyUDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.2 %
Appearance (Color)Red to yellow-brown to red-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Sensitivity(to Aluminium) Passes test