Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass Bottle
Catalog number ACR450255000
Price (MYR)
2,307.00
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,307.00
Add to cart
Chemical Identifiers
CAS4433-63-0
IUPAC Nameethylboronic acid
Molecular FormulaC2H7BO2
InChI KeyPAVZHTXVORCEHP-UHFFFAOYSA-N
SMILESCCB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0%
Appearance (Color)Colorless to white to cream
FormCrystals or powder or crystalline powder or flakes
Proton NMRConforms to structure