Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Catalog number ACR450251000
Price (MYR)
794.00
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
794.00
Add to cart
Specifications
Chemical Name or MaterialMetaphosphoric acid
CAS37267-86-0
CAS50813-16-6
PackagingGlass Bottle
Quantity100 g