2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 4781-83-3 | C4H7NS·ClH | 137.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number ACR450180010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS583-78-8
IUPAC Name2,5-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyRANCECPPZPIPNO-UHFFFAOYSA-N
SMILESOC1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals of flakes
Melting point54°C to 58°C
Appearance (Color)White to light brown
Infrared spectrumConforms
GC>=97.5 %