Check Order Status
1,4-Dioxane, 99.8%, Extra Dry, Stabilized
Element Search
Structure Search
Thermo Scientific Chemicals
1,4-Dioxane, 99.8%, Extra Dry, Stabilized
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACR450171000
100 mL
AcroSeal™ Glass Bottle
Catalog number
ACR450171000
Price (MYR)
472.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
472.00
Add to cart
Chemical Identifiers
CAS
12401-56-8
IUPAC Name
hafnium disilicon
Molecular Formula
HfSi2
InChI Key
TWRSDLOICOIGRH-UHFFFAOYSA-N
SMILES
[Si].[Si].[Hf]
View more
Specifications
Specification Sheet
Assay (unspecified)
99% min
RUO – Research Use Only