Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
CAS: 29122-68-7 | C14H22N2O3 | 266.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number ACR449550050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2363-71-5
IUPAC Namehenicosanoic acid
Molecular FormulaC21H42O2
InChI KeyCKDDRHZIAZRDBW-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Glistening crystalline powder
GC>=98.5 %