Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
CAS: 29122-68-7 | C14H22N2O3 | 266.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR449550010
Price (MYR)
539.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
539.00
Add to cart
Chemical Identifiers
CAS122-79-2
IUPAC Namephenyl acetate
Molecular FormulaC8H8O2
InChI KeyIPBVNPXQWQGGJP-UHFFFAOYSA-N
SMILESCC(=O)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow to pale brown
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4995-1.5035 @ 20?C
Assay (GC)≥96.0%