Cytidine 5'-monophosphate, 97%, from yeast
Cytidine 5'-monophosphate, 97%, from yeast
Cytidine 5'-monophosphate, 97%, from yeast
Cytidine 5'-monophosphate, 97%, from yeast
Thermo Scientific Chemicals
Cytidine 5'-monophosphate, 97%, from yeast
CAS: 63-37-6 | C9H14N3O8P | 323.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR449490050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1313-60-6
IUPAC Namedisodium dioxidanediide
Molecular FormulaNa2O2
InChI KeyPFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay contain a small quantity of discolored
Phosphate (PO4)=<5 ppm
Titration with KMnO4>=93.0 %
Heavy metals (as Pb)=<20 ppm
Sulfate (SO4)=<10 ppm