1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 L
Catalog number ACR449180250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Request bulk or custom format
Specifications
Chemical Name or Material1-Methyl-2-pyrrolidinone
Melting Point-24.0°C
CAS872-50-4
Health Hazard 1GHS Signal Word: Danger
Density1.0300g/mL
View more
Suitable for battery materials development.
RUO – Research Use Only