1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
25 L
Catalog number ACR449180050
Price (MYR)
979.00
Add to cart
Quantity:
5 L
Request bulk or custom format
Price (MYR)
979.00
Add to cart
Chemical Identifiers
CAS7021-09-2
IUPAC Name2-methoxy-2-phenylacetic acid
Molecular FormulaC9H10O3
InChI KeyDIWVBIXQCNRCFE-UHFFFAOYNA-N
SMILESCOC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Appearance (Color)Off-white to light beige
Appearance (Form)Crystalline powder
Melting point68°C to 72°C
Infrared spectrumConforms
Suitable for battery materials development.
RUO – Research Use Only