Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Diethanolamine, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethanolamine, ACS reagent, Thermo Scientific Chemicals
CAS: 111-42-2 | C4H11NO2 | 105.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ACR449062500
Price (MYR)
656.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
656.00
Add to cart
Specifications
Chemical Name or MaterialDiethanolamine
CAS111-42-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately