(-)-Epigallocatechin gallate, 95%
(-)-Epigallocatechin gallate, 95%
(-)-Epigallocatechin gallate, 95%
(-)-Epigallocatechin gallate, 95%
Thermo Scientific Chemicals
(-)-Epigallocatechin gallate, 95%
CAS: 989-51-5 | C22H18O11 | 458.37 g/mol
Have Questions?
Catalog NumberQuantity
ACR44901010010 mg
Catalog number ACR449010100
Price (MYR)
533.00
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
533.00
Add to cart
Specifications
Chemical Name or Material(-)-Epigallocatechin gallate
CAS989-51-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.