Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
Glass bottle
Catalog number ACR448532500
Price (MYR)
523.55
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
523.55
Add to cart
Specifications
Chemical Name or MaterialPoly(acrylamide), carboxyl modified
CAS62649-23-4
PackagingGlass bottle
Molecular Formula(C3H5NO)n
MDL NumberMFCD00084393