Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Thermo Scientific Chemicals
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4483910004 x 25 mLAcroSeal™ Glass Bottle
Catalog number ACR448391000
Price (MYR)
583.00
Add to cart
Quantity:
4 x 25 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
583.00
Add to cart
Specifications
Chemical Name or MaterialAcetonitrile
Melting Point-46.0°C
CAS75-05-8
ColorColorless
Health Hazard 1Danger