N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, package of 4x25ML bottles
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR448381000100 mLAcroSeal™ Glass Bottle
Catalog number ACR448381000
Price (MYR)
494.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
494.00
Add to cart
Chemical Identifiers
CAS7758-98-7
IUPAC Namecopper(2+) sulfate
Molecular FormulaCuO4S
InChI KeyARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Elemental AnalysisZinc (Zn): < 300ppm
Appearance (Color)White to pale grey
Assay from Supplier's CofA≥97.5%
Elemental AnalysisLead (Pb): < 25ppm