Mercury(II) bromide, ACS reagent
Mercury(II) bromide, ACS reagent
Mercury(II) bromide, ACS reagent
Mercury(II) bromide, ACS reagent
Thermo Scientific Chemicals
Mercury(II) bromide, ACS reagent
CAS: 7789-47-1 | Br2Hg | 360.4 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR448121000100 gGlass bottle
Catalog number ACR448121000
Price (MYR)
994.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
994.00
Add to cart
Specifications
Boiling Point322.0°C
Chemical Name or MaterialMercury(II) bromide
Melting Point236.0°C
CAS7789-47-1
Health Hazard 1GHS Signal Word: Danger