Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR448111000
Price (MYR)
514.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
514.00
Add to cart
Chemical Identifiers
CAS103-29-7
IUPAC Name(2-phenylethyl)benzene
Molecular FormulaC14H14
InChI KeyQWUWMCYKGHVNAV-UHFFFAOYSA-N
SMILESC(CC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder or fused solid or chunks
Appearance (Color)Colorless to white to pale yellow
Melting Point (clear melt)48.0-55.0?C