Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR448110025
Price (MYR)
1,930.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,930.00
Add to cart
Specifications
Chemical Name or MaterialChloroacetic acid
CAS79-11-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins