Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals
Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals
Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals
Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals
CAS: 10112-91-1 | Cl2Hg2 | 472.08 g/mol
Have Questions?
Catalog NumberQuantity
ACR447591000100 g
Catalog number ACR447591000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMercury(I) chloride
Melting Point400.0°C
CAS10112-91-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement