Gallic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Gallic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Gallic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Gallic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallic acid monohydrate, ACS reagent, Thermo Scientific Chemicals
CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR447581000
Price (MYR)
550.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
550.00
Add to cart
Specifications
Chemical Name or MaterialGallic acid monohydrate
CAS5995-86-8
Melting Point251.0°C
Molecular FormulaC7H5O5
Linear Formula(HO)3C6H2CO2H·H2O