Crystal violet, ACS reagent
Crystal violet, ACS reagent
Crystal violet, ACS reagent
Crystal violet, ACS reagent
Thermo Scientific Chemicals
Crystal violet, ACS reagent
CAS: 548-62-9 | C25H30ClN3 | 407.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass Bottle
Catalog number ACR447570500
Price (MYR)
452.00
Add to cart
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
452.00
Add to cart
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HCl>=98.0 %
Heavy metals (ICP-OES)=<0.002 %
Sulfate (SO4)=<0.05 %
Lithium carbonate=<2.0 %
View more
Indicator
RUO – Research Use Only