tert-Butyl methyl ether, ACS reagent, Thermo Scientific Chemicals
tert-Butyl methyl ether, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl methyl ether, ACS reagent, Thermo Scientific Chemicals
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Catalog NumberQuantity
ACR4474700252.5 L
Catalog number ACR447470025
Price (MYR)
683.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
683.00
Add to cart
Chemical Identifiers
CAS75-84-3
IUPAC Name2,2-dimethylpropan-1-ol
Molecular FormulaC5H12O
InChI KeyKPSSIOMAKSHJJG-UHFFFAOYSA-N
SMILESCC(C)(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or chunks or fused solid
Identification (FTIR)Conforms
Assay (GC)≥98.5%
Appearance (Color)Colorless to white
Melting Point (clear melt)51.0-58.0°C