Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
CAS: 1330-20-7 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR447240025
Price (MYR)
1,085.00
Add to cart
Quantity:
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,085.00
Add to cart
Specifications
Chemical Name or MaterialXylenes
Melting Point-34.0°C
CAS1330-20-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause