Potassium permanganate, ACS reagent, low in mercury
Potassium permanganate, ACS reagent, low in mercury
Potassium permanganate, ACS reagent, low in mercury
Potassium permanganate, ACS reagent, low in mercury
Thermo Scientific Chemicals
Potassium permanganate, ACS reagent, low in mercury
CAS: 7722-64-7 | KMnO4 | 158.032 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR447215000500 gGlass bottle
Catalog number ACR447215000
Price (MYR)
1,141.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,141.00
Add to cart
Chemical Identifiers
CAS6381-59-5
IUPAC Namepotassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
Molecular FormulaC4H12KNaO10
InChI KeyVZOPRCCTKLAGPN-ZFJVMAEJSA-L
SMILESO.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Optical Rotation+21.5 ? 1? (c=1, water)
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Oxidation reagent
RUO – Research Use Only