Potassium permanganate, ACS reagent, low in mercur
Potassium permanganate, ACS reagent, low in mercur
Thermo Scientific Chemicals
Potassium permanganate, ACS reagent, low in mercur
CAS: 7722-64-7 | KMnO4 | 158.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR447211000100 gGlass bottle
Catalog number ACR447211000
Price (MYR)
452.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
452.00
Add to cart
Chemical Identifiers
CAS3002-23-1
IUPAC Name6-methylheptane-2,4-dione
Molecular FormulaC8H14O2
InChI KeyIGMOYJSFRIASIE-UHFFFAOYSA-N
SMILESCC(C)CC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
Refractive Index1.4550-1.4590 @ 20?C