Sodium bicarbonate, 99.5%, for biochemistry
Sodium bicarbonate, 99.5%, for biochemistry
Sodium bicarbonate, 99.5%, for biochemistry
Sodium bicarbonate, 99.5%, for biochemistry
Thermo Scientific Chemicals
Sodium bicarbonate, 99.5%, for biochemistry
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR447102500250 gPlastic Bottle
Catalog number ACR447102500
Price (MYR)
370.00
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
370.00
Add to cart
Chemical Identifiers
CAS69739-34-0
IUPAC Nametert-butyldimethylsilyl trifluoromethanesulfonate
Molecular FormulaC7H15F3O3SSi
InChI KeyWLLIXJBWWFGEHT-UHFFFAOYSA-N
SMILESCC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.3850 to 1.3880 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Assay>=97.5 %
Biological buffer
RUO – Research Use Only