Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, Thermo Scientific Chemicals
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR447080010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS69-78-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point240°C to 245°C
Titration with NaOH>=98.5 %
Loss on drying=<0.5 %
Appearance (Color)Off-white to yellow