D(+)-Glucose 6-phosphate sodium salt, 98%
D(+)-Glucose 6-phosphate sodium salt, 98%
D(+)-Glucose 6-phosphate sodium salt, 98%
D(+)-Glucose 6-phosphate sodium salt, 98%
Thermo Scientific Chemicals
D(+)-Glucose 6-phosphate sodium salt, 98%
CAS: 54010-71-8 | C6H12NaO9P | 282.12 g/mol
Have Questions?
Catalog NumberQuantity
ACR4469800101 g
Catalog number ACR446980010
Price (MYR)
1,017.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,017.00
Add to cart
Chemical Identifiers
CAS4422-95-1
IUPAC Namebenzene-1,3,5-tricarbonyl trichloride
Molecular FormulaC9H3Cl3O3
InChI KeyUWCPYKQBIPYOLX-UHFFFAOYSA-N
SMILESClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid or flakes or clear liquid or viscous liquid as melt
Identification (FTIR)Conforms
Appearance (Color)Colorless to white to cream to yellow
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Melting Point (clear melt)33.0-39.0?C