Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number ACR446882500
Price (MYR)
1,643.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,643.00
Add to cart
Chemical Identifiers
CAS1121-60-4
IUPAC Namepyridine-2-carbaldehyde
Molecular FormulaC6H5NO
InChI KeyCSDSSGBPEUDDEE-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear yellow to yellow-brown
Appearance (Form)Liquid
Refractive index1.5343 to 1.5373 (20°C, 589 nm)
For HPLC
RUO – Research Use Only