Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number ACR446880500
Price (MYR)
566.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
566.00
Add to cart
Chemical Identifiers
CAS7791-20-0
IUPAC Namenickel(2+) hexahydrate dichloride
Molecular FormulaCl2H12NiO6
InChI KeyLAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofANi (Assay: ≥24.5 to ≤26.0%)
Appearance (Color)Green
Total Metal Impurities≤10ppm (≤0.0010%)
FormPowder and/or lumps or chunks
For HPLC
RUO – Research Use Only