Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number ACR446792500
Price (MYR)
394.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Chemical Identifiers
CAS1072-98-6
IUPAC Name5-chloropyridin-2-amine
Molecular FormulaC5H5ClN2
InChI KeyMAXBVGJEFDMHNV-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)133.0-141.0°C