Tetrahydrofuran, 99.5%, for analysis, stabilized with BHT, Thermo Scientific Chemicals
Tetrahydrofuran, 99.5%, for analysis, stabilized with BHT, Thermo Scientific Chemicals
Tetrahydrofuran, 99.5%, for analysis, stabilized with BHT, Thermo Scientific Chemicals
Tetrahydrofuran, 99.5%, for analysis, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofuran, 99.5%, for analysis, stabilized with BHT, Thermo Scientific Chemicals
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number ACR446400010
Price (MYR)
530.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
530.00
Add to cart
Specifications
Chemical Name or MaterialTetrahydrofuran
Melting Point-108.4°C
CAS109-99-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Suspected of causing cancer.
May form