Sodium phosphate, monobasic dihydrate, for HPLC
Sodium phosphate, monobasic dihydrate, for HPLC
Sodium phosphate, monobasic dihydrate, for HPLC
Sodium phosphate, monobasic dihydrate, for HPLC
Thermo Scientific Chemicals
Sodium phosphate, monobasic dihydrate, for HPLC
CAS: 13472-35-0 | H6NaO6P | 156.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR446222500250 gglass bottle
Catalog number ACR446222500
Price (MYR)
616.00
EA
Add to cart
Quantity:
250 g
Packaging:
glass bottle
Request bulk or custom format
Price (MYR)
616.00
EA
Add to cart
Chemical Identifiers
CAS8004-13-5
IUPAC Name1,1'-biphenyl; phenoxybenzene
Molecular FormulaC24H20O
InChI KeyMHCVCKDNQYMGEX-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=CC=C1.O(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
CommentEutectic mixture of 73% diphenyl ether and 27% biphenyl
CommentHeat transfer fluid
Appearance (Color)Clear colorless to pale yellow
pH>6.0 to <8.0