Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
CAS: 10378-23-1 | C10H12N2Na4O8·2H2O | 416.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR446085000
Price (MYR)
614.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
614.00
Add to cart
Chemical Identifiers
CAS1333-86-4
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormPowder
CommentAbsorption Stiffness Value: 30.0-34.0 ml/5g
CommentDensity: 160±20 kg/m³
CommentMoisture Content (As packed): ≤0.3%
View more
Chelation/Complexation reagent
RUO – Research Use Only