Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
CAS: 10378-23-1 | C10H12N2Na4O8·2H2O | 416.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR446081000
Price (MYR)
481.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
481.00
Add to cart
Chemical Identifiers
CAS7499-07-2
IUPAC Name4-chloro-2-methylbenzoic acid
Molecular FormulaC8H7ClO2
InChI KeyXXFKOBGFMUIWDH-UHFFFAOYSA-N
SMILESCC1=CC(Cl)=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)>97.5%
Formpowder
Assay (Silylated GC)>97.5%
Chelation/Complexation reagent
RUO – Research Use Only