Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals
CAS: 100-58-3 | C6H5BrMg | 181.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number ACR445961000
Price (MYR)
912.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
912.00
Add to cart
Chemical Identifiers
CAS97004-04-1
IUPAC Name1-(6-chloropyridin-3-yl)methanamine
Molecular FormulaC6H7ClN2
InChI KeyXPARFBOWIYMLMY-UHFFFAOYSA-N
SMILESNCC1=CN=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or pale cream to pale brown
Assay (GC)≥96.0%
Melting Point25-33?C (clear to cloudy melt)
FormPowder or Fused solid